Learn More
Meso Scale Discovery VPLEX Chemokine PNL 1HMNKit
SDP
Supplier:  Meso Scale Discovery K15047D4
Encompass
The Meso Scale Discovery V-PLEX Chemokine Panel 1 Human Kit is used to simultaneously quantify human chemokines CCL2 CCL3 CCL4 CCL5 CXCL8 CXCL9 CXCL10 and CXCL11 in serum plasma and cell culture supernatants This electrochemiluminescence (ECL) based immunoassay utilizes MSD s patented multi-array technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and diluent The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-81673
Add to Cart
Request a Quote