Learn More
Meso Scale Discovery VPLEX COVID19 COV PNL IgG
SDP
Supplier:  Meso Scale Discovery K15399U4
Encompass
The Meso Scale Discovery V-PLEX COVID-19 Coronavirus Panel 3 IgG Kit is an immunoassay that quantitatively measures IgG antibodies to SARS-CoV-2 in human serum and plasma It includes antigens for the SARS-CoV-2 spike protein receptor binding domain (RBD) and nucleocapsid protein (N) The electrochemiluminescence immunoassay is run on Meso Scale Discovery s QuickPlex SQ 120 instrument with a testing time of approximately 2 5 hours and a dynamic range up to 2500 U/mL This semi-quantitative serology test can aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection
Catalog No. 50-000-81169
Only null left
Add to Cart
Request a Quote