Learn More
Meso Scale Discovery VPLEX Cytokine PNL 2 human
SDP
Supplier:  Meso Scale Discovery K15084D4
Encompass
The Meso Scale Discovery V-PLEX Cytokine Panel 2 (human) Kit is designed to simultaneously measure IL-2 IL-4 IL-6 IL-10 IL-12p70 IL-13 IFN- and TNF- in cell culture supernatants serum plasma (EDTA heparin citrate) saliva and lysates This electrochemiluminescent assay utilizes MSD s patented SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results from a small sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG Read Buffer Diluent 2 Calibrator Blend and wash buffer
Catalog No. 50-000-81775
Add to Cart
Request a Quote