Learn More
Meso Scale Discovery VPLEX HUM Biomarker 39Plex Kit
SDP
Supplier:  Meso Scale Discovery K15209D2
Encompass
The V-PLEX Human Biomarker 40-plex Kit from Meso Scale Diagnostics is a multiplex immunoassay that allows simultaneous measurement of 40 biomarkers in human serum plasma and cell culture supernatants The kit uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The panel includes cytokines chemokines growth factors soluble receptors cardiovascular disease markers and cancer biomarkers This kit enables efficient profiling of samples for biomarker discovery and validation studies
Catalog No. 50-000-78837
Only null left
Add to Cart
Request a Quote