Learn More
Meso Scale Discovery VPLEX HUM IL17D Kit 25 PL
SDP
Supplier:  Meso Scale Discovery K151WQD4
Encompass
The V-PLEX Human IL-17D Kit from Meso Scale Discovery is a sandwich immunoassay that allows for the quantitative measurement of human interleukin 17D (IL-17D) in cell culture supernatants serum plasma and other biological fluids The assay utilizes electrochemiluminescence for high sensitivity and a broad dynamic range The kit includes a 96-well plate coated with a capture antibody specific for human IL-17D a detection antibody conjugated to an electrochemiluminescent label and calibrator diluent The assay has high sensitivity with a lower limit of detection of 0 10 pg/mL and an upper limit of quantitation of 1500 pg/mL for cell culture supernatants
Catalog No. 50-000-81894
Only null left
Add to Cart
Request a Quote