Learn More
Meso Scale Discovery VPLEX HUM Prinflm PNL 4PlxPlt
SDP
Supplier:  Meso Scale Discovery K15052G4
Encompass
The V-PLEX Plus Human ProInflam Panel I 4-plex is a multiplex immunoassay kit from Meso Scale Diagnostics for measuring 4 human proinflammatory biomarkers (IFN- IL-1 IL-6 TNF- ) in serum plasma and culture supernatants It uses electrochemiluminescence detection for high sensitivity and broad range with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies calibrators diluent wash buffer read buffer and plate seals enabling efficient analysis of inflammation biomarkers from a small sample
Catalog No. 50-000-79190
Add to Cart
Request a Quote