Learn More
Meso Scale Discovery VPLEX Metabolic PNL mouse
SDP
Supplier:  Meso Scale Discovery K15333D1
Encompass
The Meso Scale Discovery V-PLEX Metabolic Panel 1 Mouse Kit is designed to simultaneously quantify insulin leptin resistin and PAI-1 from mouse samples This 4-plex kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer wash buffer and plate seals The electrochemiluminescent immunoassay allows sensitive detection of the analytes from as little as 25 L of sample This kit provides a streamlined workflow for measuring metabolic biomarkers from mouse serum plasma and culture supernatants With high sensitivity broad dynamic range and intra-/inter-assay precision this kit enables robust metabolic profiling
Catalog No. 50-000-79377