Learn More
Meso Scale Discovery VPLEX Mouse IL27p28/IL30 Kit
SDP
Supplier:  Meso Scale Discovery K152WXD1
Encompass
The Meso Scale Discovery V-PLEX Mouse IL-27p28/IL-30 Kit is used to quantitatively measure IL-27p28 and IL-30 cytokine levels in mouse biological samples such as serum plasma and cell culture supernatants using an electrochemiluminescent detection method The kit includes a plate coated with capture antibodies detection antibodies calibrators diluent read buffer wash buffer and adhesive plate seals valid for testing 96 wells The assay has a testing time of 3 5 hours and a detection range of 48-50 000 pg/mL for IL-27p28 and 78-100 000 pg/mL for IL-30 This is an immunoassay kit ideal for studying cytokine biology and immunology in mouse models
Catalog No. 50-000-80339