Learn More
Meso Scale Discovery VPLEX Neuroinflam PNL hu1PL
SDP
Supplier:  Meso Scale Discovery K15210D1
Encompass
The Meso Scale Discovery V-PLEX Neuroinflammation Panel 1 Human Kit is a multiplex immunoassay for measuring biomarkers related to neuroinflammation and neurodegeneration It includes a 96-well plate detection antibodies calibrators controls diluent wash buffer read buffer and plate seals It measures human amyloid beta 1-42 amyloid beta 1-40 total tau phosphorylated tau C-reactive protein S100B and GFAP in serum plasma CSF and cell culture supernatants aiding research on Alzheimer s brain injury stroke and other neuroinflammatory conditions using electrochemiluminescence detection
Catalog No. 50-000-78448
Only null left
Add to Cart
Request a Quote