Learn More
Meso Scale Discovery VPLEX Neuroinflammation PNL hu
SDP
Supplier:  Meso Scale Discovery K15210D4
Encompass
The Meso Scale Discovery V-PLEX Neuroinflammation Panel 1 Human Kit is a multiplex immunoassay that simultaneously measures biomarkers related to neuroinflammation and neurodegeneration This kit includes a 96-well plate detection antibodies calibrators diluents wash buffers readout buffers and plate seals needed to quantitatively measure human amyloid beta 1-42 amyloid beta 1-40 total tau phosphorylated tau C-reactive protein and vascular cell adhesion molecule 1 from serum plasma and CSF samples This kit utilizes MSD s electrochemiluminescent detection technology for high sensitivity and a wide dynamic range
Catalog No. 50-000-81349
Only null left
Add to Cart
Request a Quote