Learn More
Meso Scale Discovery VPLEX NHP TNFSY Kit 25Plate
SDP
Supplier:  Meso Scale Discovery K156RGD4
Encompass
The V-PLEX NHP TNF-beta Kit from Meso Scale Discovery is a highly sensitive immunoassay that quantitatively measures natural human TNF-beta in serum plasma and cell culture supernatants The kit utilizes electrochemiluminescent detection technology and provides accurate results in just 3 5 hours The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator solutions for generating a standard curve The assay has a sensitivity of 0 04 pg/mL and requires just 25 L of sample This TNF-beta kit enables fast reliable quantification of natural human TNF-beta for research applications
Catalog No. 50-000-78550
Add to Cart
Request a Quote