Learn More
Meso Scale Discovery VPLEX OPV PNL IgG
SDP
Supplier:  Meso Scale Discovery K15688U2
Encompass
The Meso Scale Discovery V-PLEX Orthopoxvirus Panel 1 IgG Kit is an electrochemiluminescence immunoassay that detects IgG antibodies to vaccinia virus monkeypox virus cowpox virus and ectromelia virus in serum and plasma samples This multiplex assay uses patterned arrays and MSD s proprietary SULFO-TAG labels to provide quantitative results The kit includes a 96-well plate coated with antigens detection antibody wash buffer read buffer calibrator and controls This assay allows high-throughput sensitive and reproducible detection of orthopoxvirus IgG antibodies
Catalog No. 50-000-81009
Add to Cart
Request a Quote