Learn More
Meso Scale Discovery VPLEX Plus Ab Peptide PNL 6E10
SDP
Supplier:  Meso Scale Discovery K15200G1
Encompass
The Meso Scale Discovery V-PLEX Plus Abeta Peptide Panel 1 (6E10) Kit is a multiplex immunoassay that measures Abeta38 Abeta40 and Abeta42 in human CSF and plasma samples It uses electrochemiluminescence detection technology and provides quantitative results in as little as 3 5 hours The kit includes a 96-well plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance It allows sensitive reproducible and accurate measurement of Abeta peptides that are important biomarkers of Alzheimer s disease
Catalog No. 50-000-78928