Learn More
Meso Scale Discovery VPLEX Plus Ab Peptide PNL 6E10
SDP
Supplier:  Meso Scale Discovery K15200G4
Encompass
The Meso Scale Discovery V-PLEX Plus Abeta Peptide Panel 1 (6E10) Kit is used to quantitatively measure Abeta38 Abeta40 and Abeta42 in human CSF and plasma samples This electrochemiluminescence (ECL) immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results The kit includes a 96-well plate coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and controls The assay has a typical lower limit of quantification of 1 95 pg/mL for Abeta38 1 56 pg/mL for Abeta40 and 0 61 pg/mL for Abeta42
Catalog No. 50-000-79870
Add to Cart
Request a Quote