Learn More
Meso Scale Discovery VPLEX Plus Ab40 Peptide 4G8
SDP
Supplier:  Meso Scale Discovery K150SJG1
Encompass
The Meso Scale Discovery V-PLEX Plus Abeta40 Peptide Kit quantitatively measures Abeta40 in human serum plasma CSF and cell culture supernatants This electrochemiluminescence (ECL) immunoassay uses MSD s SULFO-TAG labels and multi-array plate technology for high sensitivity and low sample volume The kit includes a pre-coated plate detection antibody with ruthenium label calibrators and necessary reagents Results are available in 3 5 hours using an MSD ECL detection instrument aiding Alzheimer s disease research
Catalog No. 50-000-81778