Learn More
Meso Scale Discovery VPLEX Plus Ab40 Peptide 6E10
SDP
Supplier:  Meso Scale Discovery K150SKG1
Encompass
The Meso Scale Discovery V-PLEX Plus A 40 Peptide Kit is an immunoassay that quantitatively measures A 40 levels in human CSF and plasma samples It uses electrochemiluminescence detection technology and provides high sensitivity a broad dynamic range and requires low sample volumes The kit includes a plate coated with a monoclonal antibody to the N-terminus of A 40 a detection antibody targeting the C-terminus calibrators controls buffers reagents and plates for 96 tests It can detect A 40 levels between 1 53-1500 pg/mL in CSF and 0 244-300 pg/mL in plasma The assay takes 3 5 hours to complete
Catalog No. 50-000-78323