Learn More
Meso Scale Discovery VPLEX Plus Chemokine PNL NHP
SDP
Supplier:  Meso Scale Discovery K15055G1
Encompass
The Meso Scale Discovery V-PLEX Plus Chemokine Panel 1 NHP Kit is a multiplex immunoassay for measuring 10 non-human primate chemokines in serum plasma and culture supernatants It includes a 96-well plate with capture antibodies detection antibodies calibrators controls diluent read buffer wash buffer and adhesive plate seals Sensitivity is 0 3-43 pg/mL requiring 25 L of sample Analytes CCL2/MCP-1 CCL3/MIP-1 CCL4/MIP-1 CCL5/RANTES CXCL8/IL-8 CXCL9/MIG CXCL10/IP-10 CXCL11/I-TAC CXCL12/SDF-1 CX3CL1/Fractalkine This assay quantifies chemokine levels efficiently
Catalog No. 50-000-80522