Learn More
Meso Scale Discovery VPLEX Plus Chemokine PNL NHP
SDP
Supplier:  Meso Scale Discovery K15055G2
Encompass
The Meso Scale Discovery V-PLEX Plus Chemokine Panel 1 NHP Kit is a multiplex immunoassay kit for the quantitative measurement of 5 chemokines in non-human primate samples The kit includes a 5-plex panel of analyte-specific antibodies and calibrators for the detection of CCL2 CCL3 CCL4 CXCL8 and CXCL10 It utilizes electrochemiluminescent detection technology on Meso Scale Discovery s QuickPlex SQ 120 instrument The kit contains reagents for 5 x 96-well plates and can assay up to 80 samples in duplicate It is intended for research use only
Catalog No. 50-000-80523
Add to Cart
Request a Quote