Learn More
Meso Scale Discovery VPLEX Plus HUM IL12 p70 Kit
SDP
Supplier:  Meso Scale Discovery K151QVG1
Encompass
The Meso Scale Discovery V-PLEX Plus Human IL-12p70 Kit is an electrochemiluminescent immunoassay for measuring human interleukin 12 subunit p70 (IL-12p70) in various biological fluids It includes a 96-well plate with a capture antibody a detection antibody with an electrochemiluminescent label calibrators diluents wash buffers and necessary reagents The kit detects 3 2-10 000 pg/mL and shows no cross-reactivity with other cytokines enabling sensitive and accurate IL-12p70 quantification in about 3 5 hours
Catalog No. 50-000-81265