Learn More
Meso Scale Discovery VPLEX Plus HUM TSLP
SDP
Supplier:  Meso Scale Discovery K151XEG4
Encompass
The V-PLEX Plus Human TSLP Kit is a multiplex immunoassay kit from Meso Scale Diagnostics that allows for the simultaneous quantitative measurement of human Thymic Stromal Lymphopoietin (TSLP) in serum plasma and cell culture supernatants The electrochemiluminescent detection method provides high sensitivity and a broad dynamic range This research tool enables the study of TSLP and its role in allergic diseases and asthma The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer wash buffer and plate seals
Catalog No. 50-000-81831
Only null left
Add to Cart
Request a Quote