Learn More
Meso Scale Discovery VPLEX Plus Metabolic PNL mouse
SDP
Supplier:  Meso Scale Discovery K15333G4
Encompass
The V-PLEX Plus Metabolic Panel 1 Mouse Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows simultaneous quantitative measurement of 10 metabolic biomarkers in mouse serum plasma and tissue culture media samples The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls diluent read buffer wash buffer and plate seals The assay has a short 2 5 hour protocol and a broad dynamic range to accurately quantify insulin leptin GLP-1 PAI-1 resistin glucagon GIP MCP-1 amylin and C-peptide from a single small sample volume This kit enables efficient metabolic profiling of mouse models
Catalog No. 50-000-81306
Only null left
Add to Cart
Request a Quote