Learn More
Meso Scale Discovery VPLEX Plus Viral PNL 2HMNKit
SDP
Supplier:  Meso Scale Discovery K15346G2
Encompass
The Meso Scale Discovery V-PLEX Plus Viral Panel 2 Human Kit is a multiplex immunoassay that simultaneously detects antibodies to SARS-CoV-2 Influenza A Influenza B and RSV in human serum and plasma This assay kit includes a 96-well plate coated with antigens for the viruses detection antibodies assay buffers wash buffers calibrators and controls The electrochemiluminescence technology allows for high sensitivity and a broad dynamic range This kit can help track immune responses to viral infections and vaccines with results in just a few hours
Catalog No. 50-000-79805
Only null left
Add to Cart
Request a Quote