Learn More
Meso Scale Discovery VPLEX Respiratory PNL 1 IgA
SDP
Supplier:  Meso Scale Discovery K15367U4
Encompass
The Meso Scale Diagnostics COVID-19 Respiratory Panel 1 IgA is an in vitro diagnostic test kit that detects IgA antibodies to SARS-CoV-2 in human serum and plasma This qualitative immunoassay uses electroluminescence technology on microplates coated with SARS-CoV-2 antigens Results are available in under 3 hours This product requires the MSD Discovery Workbench or QuickPlex SQ 120 instrument and is intended for use by professional laboratories and healthcare workers as an aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection
Catalog No. 50-000-80350
Only null left
Add to Cart
Request a Quote