Learn More
Meso Scale Discovery VPLEX SARSCoV2 384 PNL 1 IgG
SDP
Supplier:  Meso Scale Discovery K25392U4
Encompass
The V-PLEX Human Cytokine 30-Plex Kit is an immunoassay for measuring 30 human cytokines and chemokines in serum plasma and culture supernatants It uses electrochemiluminescence detection and patterned arrays for high sensitivity broad dynamic range and precision The kit includes a 96-well plate with capture antibodies detection antibodies standards controls buffers substrate and an MSD instrument Key analytes include IL-2 IL-4 IL-6 IL-10 IL-17A/F IFN-gamma TNF-alpha and VEGF enabling efficient cytokine profiling
Catalog No. 50-000-81290
Add to Cart
Request a Quote