Learn More
Meso Scale Discovery VPLEX SARSCoV2 384 PNL ACE2
SDP
Supplier:  Meso Scale Discovery K25395U2
Encompass
The Meso Scale Discovery V-PLEX Human IL-6 Kit is a highly sensitive and quantitative assay for measuring human interleukin 6 (IL-6) in serum plasma and cell culture supernatants This electrochemiluminescent immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to enable simultaneous detection of IL-6 down to 0 05 pg/mL with an upper limit of 10 000 pg/mL The kit includes a 96-well plate pre-coated with capture antibody detection antibody conjugated to ruthenium calibrators and reagents required to perform the assay It provides a fast and reliable way to quantify IL-6 levels with high sensitivity and wide dynamic range
Catalog No. 50-000-79464
Add to Cart
Request a Quote