Learn More
Meso Scale Discovery VPLEX SARSCoV2 384 PNL IgG
SDP
Supplier:  Meso Scale Discovery K25392U2
Encompass
The Meso Scale Discovery SARS-CoV-2 384 Panel 1 IgG is an immunoassay panel that tests for IgG antibodies to SARS-CoV-2 in human serum and plasma It uses electrochemiluminescence detection technology and provides quantitative results for IgG antibodies to the SARS-CoV-2 spike protein receptor binding domain and nucleocapsid protein The panel allows high-throughput testing with 384-well plates It has high sensitivity and specificity and has received FDA Emergency Use Authorization The panel aids in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection
Catalog No. 50-000-81289
Add to Cart
Request a Quote