Learn More
Meso Scale Discovery VPLEX SARSCoV2 KVSpike PNL IgG
SDP
Supplier:  Meso Scale Discovery K15651U4
Encompass
The Meso Scale Discovery V-PLEX SARS-CoV-2 Key Variant Spike Panel 1 IgG Kit detects IgG antibodies to the spike protein of SARS-CoV-2 and its key variants in human serum and plasma This assay measures antibodies to the spike protein of the original Wuhan strain and the Alpha Beta Gamma Delta and Omicron variants It uses electrochemiluminescent detection for high sensitivity The kit includes a plate with recombinant spike proteins detection antibodies calibrators controls diluent read buffer wash buffer and plate seals with a throughput of 30 samples in duplicates per plate and a 25 L sample volume
Catalog No. 50-000-80961
Add to Cart
Request a Quote