Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 1 IgA
SDP
Supplier:  Meso Scale Discovery K15361U4
Encompass
The Meso Scale Discovery SARS-CoV-2 Panel 1 IgA is an electrochemiluminescence immunoassay for the qualitative detection of IgA antibodies to SARS-CoV-2 in human serum and plasma This assay can aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection The panel allows for the testing of up to 30 samples in duplicate in less than 3 hours The assay has high sensitivity and specificity and has received FDA Emergency Use Authorization This product would be useful for seroprevalence studies and research applications
Catalog No. 50-000-79373
Only null left
Add to Cart
Request a Quote