Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 2 IgG Kit
SDP
Supplier:  Meso Scale Discovery K15383U2
Encompass
The Meso Scale Discovery SARS-CoV-2 Panel 2 IgG is an in vitro diagnostic test kit that detects IgG antibodies to SARS-CoV-2 in human serum and plasma It uses electrochemiluminescence technology and provides quantitative results for IgG antibodies to the receptor binding domain and nucleocapsid protein of SARS-CoV-2 The test has high sensitivity and specificity and is intended to aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection The kit includes a 96-well plate coated with antigens detection antibody read buffer sample diluent calibrators and controls
Catalog No. 50-000-81968