Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 2 MSE IgA
SDP
Supplier:  Meso Scale Discovery K15477U4
Encompass
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 2 Mouse IgA Kit is an electrochemiluminescence immunoassay that quantitatively measures IgA antibodies to SARS-CoV-2 in mouse serum and plasma samples The assay uses a multiplex format to measure antibodies to the SARS-CoV-2 spike protein receptor binding domain and nucleocapsid protein The kit includes a plate coated with antigens detection antibodies calibrators controls diluent and wash buffer The assay has a typical lower limit of detection of 1 95 units/mL and an upper limit of 500 units/mL It requires 50 L of sample and can process 96 tests in under 3 hours
Catalog No. 50-000-78284
Only null left
Add to Cart
Request a Quote