Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 24 IgA
SDP
Supplier:  Meso Scale Discovery K15577U4
Encompass
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 2 IgA Kit is an electrochemiluminescence immunoassay that quantitatively measures IgA antibodies to SARS-CoV-2 in human serum and plasma This assay uses a multiplex format to measure antibodies to the SARS-CoV-2 spike and nucleocapsid proteins The kit includes a 96-well plate coated with antigens detection antibodies calibrators controls diluent and other reagents required to perform the assay Results are obtained in under 3 hours using an MSD instrument This assay can aid in understanding the adaptive immune response to SARS-CoV-2 infection
Catalog No. 50-000-78234
Only null left
Add to Cart
Request a Quote