Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 24 MSE IgG
SDP
Supplier:  Meso Scale Discovery K15579U4
Encompass
The Mesoscale K15579U-4 is a high-performance thermoelectric module that can be used for cooling or heating applications It measures 40 x 40 x 3 4mm and provides a maximum heat pumping capacity of 15 6W The module operates on 15V and can achieve a maximum temperature difference of 68 C It utilizes Bismuth Telluride-based alloys as n-type and p-type semiconductors The module has a no-load voltage of 3 66V and internal resistance of 1 15 ohms It can withstand peak operating temperatures between -40 C to 125 C The K15579U-4 module needs to be paired with a heat sink and fan for optimal performance Key features include high reliability stability and energy efficiency
Catalog No. 50-000-82321
Only null left
Add to Cart
Request a Quote