Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 27 MSE IgG
SDP
Supplier:  Meso Scale Discovery K15610U2
Encompass
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 2 Mouse IgG Kit is an electrochemiluminescence immunoassay that detects IgG antibodies to SARS-CoV-2 in mouse serum and plasma samples This assay uses a multiplex format to measure antibodies specific for the SARS-CoV-2 spike and nucleocapsid proteins The kit includes a plate coated with antigens detection antibodies calibrators controls diluent and wash buffer Results are obtained in under 3 hours using an MSD instrument This assay allows for high-throughput quantitative measurement of SARS-CoV-2 IgG antibodies
Catalog No. 50-000-78595
Only null left
Add to Cart
Request a Quote