Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 33 IgG Kit
SDP
Supplier:  Meso Scale Discovery K15676U2
Encompass
The V-PLEX SARS-CoV-2 Panel 33 IgG Kit from Meso Scale Diagnostics is an immunoassay that uses electrochemiluminescence technology to detect IgG antibodies against 33 antigens from SARS-CoV-2 in human serum and plasma This qualitative assay can aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection The kit includes a 96-well plate coated with SARS-CoV-2 antigens detection antibodies calibrators controls diluent wash buffer read buffer and plate seals Results are analyzed using MSD s imaging systems This assay has high sensitivity and specificity and can be run in approximately 5 hours
Catalog No. 50-000-80894
Only null left
Add to Cart
Request a Quote