Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 34 IgG Kit
SDP
Supplier:  Meso Scale Discovery K15690U2
Encompass
The V-PLEX SARS-CoV-2 Panel 1 IgG Kit is a serology test kit that detects IgG antibodies to SARS-CoV-2 in human serum and plasma It uses Meso Scale Discovery s electrochemiluminescence technology and provides quantitative results for IgG antibodies to the SARS-CoV-2 spike and nucleocapsid proteins The test has high sensitivity and specificity and is intended for use as an aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection The kit includes reagents for 25 microplate wells and can process up to 21 samples in duplicate
Catalog No. 50-000-80867
Only null left
Add to Cart
Request a Quote