Learn More
Meso Scale Discovery VPLEX Viral PNL 1 human Kit
SDP
Supplier:  Meso Scale Discovery K15345D4
Encompass
The Meso Scale Discovery V-PLEX Viral Panel 1 Human Kit is a multiplex immunoassay that simultaneously detects antibodies for SARS-CoV-2 Influenza A Influenza B and RSV in human serum and plasma This assay utilizes electrochemiluminescence detection technology and provides quantitative results in under 3 hours The kit includes a 96-well plate coated with antigens detection antibodies calibrators controls wash buffer read buffer and plate seals This assay allows high-throughput sensitive and accurate serological testing for common respiratory viruses
Catalog No. 50-000-81519
Only null left
Add to Cart
Request a Quote