Learn More
Meso Scale Discovery VPLEXus NHP IL12/IL23p405
SDP
Supplier:  Meso Scale Discovery K156RJG2
Encompass
Meso Scale Diagnostics V-PLEX Plus Cytokine Panel 1 (human) Kit enables the quantitative measurement of 10 cytokines and chemokines (IFN- IL-10 IL-12p70 IL-13 IL-1 IL-2 IL-4 IL-6 IL-8 TNF- ) from one sample This multiplex immunoassay uses electrochemiluminescence detection on the QuickPlex SQ120 instrument The kit includes a 96-well plate with capture antibodies detection antibodies with an electrochemiluminescent label and calibrator solutions allowing high sensitivity cytokine quantification from human serum plasma culture supernatants and other samples
Catalog No. 50-000-80824