Learn More
Cayman Chemical VPMp15trIfluoroacetAT sal 5mg
SDP
Supplier:  Cayman Chemical 374705 MG
Encompass_Preferred
A peptide ADGRG2 agonist; induces cAMP accumulation in HEK293 cells expressing human ADGRG2 (EC50 = 1.41 µM)
Catalog No. 50-291-7358