Learn More
Selleck Chemical LLC VPS34-IN1 5mg 1383716-33-3 Vps34-IN-1
SDP
Supplier:  Selleck Chemical LLC S79805MG
Encompass_Preferred
Vps34-IN1 is a potent and highly selective Vps34 inhibitor with IC50 of 25 nM invitro,which does not significantly inhibit the isoforms of class I as well as class II PI3Ks. Vps34-IN1 modulates autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5506