Learn More
ATCC ATCC VR-415 Modoc Virus (Strain M 544), lyophilized
Supplier: ATCC VR415
Modoc virus (strain M 544) is a lyophilized viral isolate originally recovered from Peromyscus maniculatus (mouse) mammary gland in California and deposited in 1958. The agent produces hemagglutinin and exhibits one-way cross-immunity with Rio Bravo; no arthropod vector has been demonstrated. Requests for this agent require a signed statement assuming responsibility for laboratory handling.
- Product format: Freeze-dried.
- Recommended storage: -70°C or colder.
- Source organism: Peromyscus maniculatus (mouse), lactating mammary gland.
- Strain: M 544.
- Deposited: 1958.
- Hemagglutinin production: Yes.
- Mycoplasma contamination: Not detected.
- Handling requirements: Signed statement assuming handling responsibility required.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.