Learn More
Selleck Chemical LLC VR23 5mg 1624602-30-7
SDP
Supplier:  Selleck Chemical LLC S79335MG
Encompass_Preferred
VR23 is a potent proteasome inhibitor with IC50 of 1 nM, 50-100 nM, and 3 ?M for trypsin-like proteasomes, chymotrypsin-like proteasomes, and caspase-like proteasomes, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5435