Learn More
Medchemexpress LLC HY-18741 5mg Medchemexpress, VR23 CAS:1624602-30-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY187415MG
Encompass_Preferred
Medchemexpress, HY-18741 5mg VR23 CAS:1624602-30-7 VR23 is a small molecule that potently inhibits the activities of trypsin-like proteasomes (IC50=1 nM), chymotrypsin-like proteasomes (IC50=50-100 nM), and caspase-like proteasomes (IC50=3 μM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2081