Learn More
Accuflow Systems Saint-Gobain Versilon 2001 Tubing, Plasticizer-Free, 1/4 in ID, 50 ft
Supplier:  Accuflow Systems AE300017
Encompass_Preferred
Versilon 2001 tubing is a plasticizer-free, chemical-resistant thermoplastic elastomer tubing designed for pump and fluid-transfer applications requiring clear visual flow monitoring and suitability for food-contact use where low extractables are important.
  • Plasticizer and oil free for low extractables and reduced leachables.
  • Clear tubing permits visual inspection of flow.
  • Inner diameter 1/4 in, outer diameter 3/8 in, wall thickness 1/16 in.
  • 50 ft length for continuous runs or cut-to-length installation.
  • Chemical resistant to a wide range of fluids.
  • Meets FDA 21 CFR 177.2600 for food contact suitability.
  • Flexible for use with peristaltic and other pump types.
Catalog No. 50-236-5855