Learn More
Medchemexpress LLC VRT-532 | 50MG
SDP
Supplier:  Medchemexpress LLC HY11472550MG
Encompass_Preferred
VRT-532 (CFpot-532) is a potent is a potent CFTR modulator VRT-532 enhances channel activity in G551D-CFTR and intrinsic ATPase activity of G551D-CFTR VRT-532 has the potential for the research of cystic fibrosis[1][2]
Catalog No. 50-002-29640