Learn More
Medchemexpress LLC VRT-532 5mg | 38214-71-0 | 5 MG

Supplier: Medchemexpress LLC HY1147255MG
A small-molecule CFTR modulator that enhances G551D-CFTR channel activity and intrinsic ATPase activity; supplied for research use in cystic fibrosis studies.
- Potent CFTR modulator activity relevant to cystic fibrosis research.
- High purity (99.0%).
- Molecular weight 250.30 g/mol; formula C16H14N2O.
- Available in small solid quantities and DMSO solutions.
- Recommended solvent: DMSO; powder storage at -20°C, solutions at -80°C for long-term.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.