Learn More
Selleck Chemical LLC VS-4718
SDP
Supplier:  Selleck Chemical LLC S76531G
Encompass_Preferred
VS-4718 (PND-1186) is a reversible and selective FAK inhibitor with IC50 of 1 5 nM PND-1186 selectively promotes tumor cell apoptosis Phase 1
Catalog No. 50-000-00121