Learn More
Chemie Tek VS-4718, 1061353-68-1, MFCD28125506, 10 mg
SDP
Supplier:  Chemie Tek CTVS471810MG
Encompass_Preferred
Formula: C25H26F3N5O3 • 0.8H2O, MW: 515.9, Synonym: PND-1186. VS-4718 (PND-1186) is a potent, selective and orally bioavailable FAK inhibitor with IC50 at low nM range.
Catalog No. NC0987545