Learn More
Medchemexpress LLC VT-105 | 2417718-38-6 | 99.93% | 421.41 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1636441MG
Encompass_Preferred
VT-105 is a TEAD inhibitor and an analog of VT104. It works by binding to the central hydrophobic pocket of the TEAD3 protein. This compound can be used in research related to NF2-deficient malignant mesothelioma. The product appears as a white to off-white solid.
  • TEAD inhibitor
  • Analog of VT104
  • Binds to the central hydrophobic pocket of the TEAD3 protein
  • Used in research of NF2-deficient malignant mesothelioma
  • White to off-white solid appearance
Catalog No. 50-003-94738