Learn More
Medchemexpress LLC Vt-1598 | 2089320-99-8 | 99.9% | 584.52 g·mol⁻¹ | C31H20F4N6O2 | 100MG
SDP
Supplier:  Medchemexpress LLC HY123777100MG
Encompass_Preferred
VT-1598 is a tetrazole-based, orally active, selective inhibitor of fungal sterol 14α-demethylase (CYP51) used as a research compound to evaluate antifungal activity. The compound has formula C31H20F4N6O2, molecular weight 584.52 g·mol⁻¹, CAS 2089320-99-8, and is supplied as a powder at high purity for laboratory studies.
Catalog No. 50-002-36802