Learn More
MSC Vermont Gage Pin Gage, Class ZZ, Tool Steel, 22.05 mm, 1 ea.
Supplier:  MSC 68113109
Encompass
This Class ZZ Plus pin gage is designed for precision inspection tasks, including go-inspection, checking the calibration of micrometers, and verifying verniers or other measuring devices. Manufactured from durable tool steel, this gage provides high accuracy and reliable performance in metrology applications.
  • Constructed from tool steel for enhanced durability
  • Features a 22.05 mm diameter with a plus tolerance direction
  • Includes a certificate of accuracy with NIST traceability
  • Designed to meet ASME B89.1 standards
  • Reversible style allows for extended wear life
Catalog No. 50-005-11938